missing translation for 'onlineSavingsMsg'
Learn More

Tenascin-X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13595-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881884

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is raised against human TNXB and detects a band about 75 kDa, which probably represent a proteolytic cleaved form of human TNXB.

Tenascin-X (TNXB) is a extracellular matrix glycoprotein predomitly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.
Specifications

Specifications

Tenascin-X
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22105
Hexabrachion like protein, HXBL, Tenascin X, tenascin XB, TENX, TN X, TNX, TNXB, TNXB1, TNXB2, TNXBS, XB, XBS
Rabbit
IgG
20 μL
-20°C
Primary
415089, 7148, 81877
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Unconjugated
TNXB
Liquid
Tnxb
Tenascin-X Fusion Protein Ag4501
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers