missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin-X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Tenascin-X |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881884
|
Proteintech
13595-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16871884
|
Proteintech
13595-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is raised against human TNXB and detects a band about 75 kDa, which probably represent a proteolytic cleaved form of human TNXB.
Tenascin-X (TNXB) is a extracellular matrix glycoprotein predomitly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.Specifications
Tenascin-X | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P22105 | |
415089, 7148, 81877 | |
Tenascin-X Fusion Protein Ag4501 | |
Primary | |
-20°C | |
TNXB |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Hexabrachion like protein, HXBL, Tenascin X, tenascin XB, TENX, TN X, TNX, TNXB, TNXB1, TNXB2, TNXBS, XB, XBS | |
Tnxb | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |