Läs mer
Teneligliptin Carboxylic Acid, TRC
Produktkod. 30422518
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30422518 25 mg
1 options
Produktkod. 30422518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT01833525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Teneligliptin Carboxylic Acid
CAS 2407632-38-4
Molekylformel C19H25N5O2
Kvantitet 25 mg
Synonym (2S,4S)-4-(4-(3-Methyl-1-phenyl-1H-pyrazol-5-yl)piperazin-1-yl)pyrrolidine-2-carboxylic Acid, Teneligliptin Impurity E
InChI formel InChI=1S/C19H25N5O2/c1-14-11-18(24(21-14)15-5-3-2-4-6-15)23-9-7-22(8-10-23)16-12-17(19(25)26)20-13-16/h2-6,11,16-17,20H,7-10,12-13H2,1H3,(H,25,26)/t16-,17-/m0/s1
LEDER CC1=NN(C2=CC=CC=C2)C(N3CCN([C@H]4C[C@H](NC4)C(O)=O)CC3)=C1
IUPAC-namn (2S,4S)-4-(4-(3-methyl-1-phenyl-1H-pyrazol-5-yl)piperazin-1-yl)pyrrolidine-2-carboxylic acid
Molekylvikt (g/mol) 355.44
Formel vikt 355.2