Läs mer
Tenofovir Alafenamide, TRC
Produktkod. 30012234
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012234 100 mg
30009244 25 mg
30004441 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012234 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT018555100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 379270-37-8
Molekylformel C21 H29 N6 O5 P
Molekylvikt (g/mol) 476.47
Synonym N-[(S)-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phenoxyphosphinyl]-L-alanine 1-Methylethyl Ester, GS 7340, Tenofovir Alafenamide
IUPAC-namn propan-2-yl (2S)-2-[[[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-phenoxyphosphoryl]amino]propanoate
LEDER CC(C)OC(=O)[C@H](C)N[P@@](=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)Oc3ccccc3
Specifikationer
Kemiskt namn eller material Tenofovir Alafenamide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C21H29N6O5P/c1-14(2)31-21(28)16(4)26-33(29,32-17-8-6-5-7-9-17)13-30-15(3)10-27-12-25-18-19(22)23-11-24-20(18)27/h5-9,11-12,14-16H,10,13H2,1-4H3,(H,26,29)(H2,22,23,24)/t15-,16+,33-/m1/s1
Formel vikt 476.1937
Procent renhet >95%