Läs mer
Tenofovir Bis(L-alanine Isopropyl Ester) Amide, TRC
Produktkod. 30424065
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424065 50 mg
30422803 100 mg
30416506 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT01856550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1883563-86-7
Molekylformel C21H36N7O6P
Molekylvikt (g/mol) 513.53
Synonym N,N′-[[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphinylidene]bis-L-Alanine 1,1′-Bis(1-methylethyl) Ester
IUPAC-namn propan-2-yl (2S)-2-[[[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-[[(2S)-1-oxo-1-propan-2-yloxypropan-2-yl]amino]phosphoryl]amino]propanoate
LEDER CC(C)OC(=O)[C@H](C)NP(=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)N[C@@H](C)C(=O)OC(C)C
Specifikationer
Kemiskt namn eller material Tenofovir Bis(L-alanine Isopropyl Ester) Amide
InChI formel InChI=1S/C21H36N7O6P/c1-12(2)33-20(29)15(6)26-35(31,27-16(7)21(30)34-13(3)4)11-32-14(5)8-28-10-25-17-18(22)23-9-24-19(17)28/h9-10,12-16H,8,11H2,1-7H3,(H2,22,23,24)(H2,26,27,31)/t14-,15+,16+/m1/s1
Kvantitet 50 mg
Formel vikt 513.2465