Läs mer
Tenofovir Diphosphate Triethylamine Salt (mixture of diastereomers), TRC
Produktkod. 30422902
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30422902 1 mg
1 options
Produktkod. 30422902 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0185251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tenofovir diphosphate triethylamine salt
CAS 2122333-63-3
Anteckningar om renhetsgrad HPLC
Molekylformel C9 H16 N5 O10 P3 . C6 H15 N
Kvantitet 1 mg
Synonym Diphosphoric Acid Triethylamine Salt ','Anhydride with [[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid Triethylamine Salt, PMPApp Triethylamine Salt
InChI formel InChI=1S/C9H16N5O10P3.C6H15N/c1-6(2-14-4-13-7-8(10)11-3-12-9(7)14)22-5-25(15,16)23-27(20,21)24-26(17,18)19;1-4-7(5-2)6-3/h3-4,6H,2,5H2,1H3,(H,15,16)(H,20,21)(H2,10,11,12)(H2,17,18,19);4-6H2,1-3H3/t6-;/m1./s1
LEDER CCN(CC)CC.C[C@H](Cn1cnc2c(N)ncnc12)OCP(=O)(O)OP(=O)(O)OP(=O)(O)O
IUPAC-namn [(1R)-2-(6-aminopurin-9-yl)-1-methyl-ethoxy]methyl-[hydroxy(phosphonooxy)phosphoryl]oxy-phosphinic acid;x-triethylamine
Molekylvikt (g/mol) 447.17
Formel vikt 447.011