Läs mer
Tenofovir Disoproxil Isopropoxycarbonyl, TRC
Produktkod. 17914525
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17914525 1 mg
17993685 5 mg
17960665 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17914525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0185201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1244022-54-5
Molekylformel C23 H36 N5 O12 P
Molekylvikt (g/mol) 605.53
Synonym 2,4,6,8-Tetraoxa-5-phosphanonanedioic acid, 5-[[(1R)-1-methyl-2-[6-[[(1-methylethoxy)carbonyl]amino]-9H-purin-9-yl]ethoxy]methyl]-, 1,9-bis(1-methylethyl) ester, 5-oxide, [[(2R)-1-[6-(Isopropyloxycarbonylamino)-9H-purin-9-yl]propan-2-yloxy]methyl(isopropyloxycarbonyloxymethoxy)phosphoryl]oxymethyl Isopropyl Carbonate
IUPAC-namn propan-2-yl [[(2R)-1-[6-(propan-2-yloxycarbonylamino)purin-9-yl]propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl carbonate
LEDER CC(C)OC(=O)Nc1ncnc2c1ncn2C[C@@H](C)OCP(=O)(OCOC(=O)OC(C)C)OCOC(=O)OC(C)C
Specifikationer
Kemiskt namn eller material Tenofovir Disoproxil Isopropoxycarbonyl
Kvantitet 1 mg
InChI formel InChI=1S/C23H36N5O12P/c1-14(2)38-21(29)27-19-18-20(25-9-24-19)28(10-26-18)8-17(7)35-13-41(32,36-11-33-22(30)39-15(3)4)37-12-34-23(31)40-16(5)6/h9-10,14-17H,8,11-13H2,1-7H3,(H,24,25,27,29)/t17-/m1/s1
Formel vikt 605.2098