Läs mer
Tenuazonic Acid Copper Salt, TRC
Produktkod. 30008485
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008485 1 mg
17909849 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT0196201MG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 12427-40-6
Molekylformel 2 C10 H14 N O3 . Cu
Molekylvikt (g/mol) 455.99
Synonym L-3-Acetyl-5-sec-butyl-4-hydroxy-3-pyrrolin-2-one Copper Salt, (5S)-3-Acetyl-1,5-dihydro-4-hydroxy-5-[(1S)-1-methylpropyl]-2H-pyrrol-2-one Copper Salt, AAC-toxin Copper Salt, L-Tenuazonic Acid Copper Salt
IUPAC-namn copper;(2S)-4-acetyl-2-[(1S)-1-methylpropyl]-5-oxo-1,2-dihydropyrrol-3-olate
LEDER [Cu+2].CC[C@H](C)[C@@H]1NC(=O)C(=C1[O-])C(=O)C.CC[C@H](C)[C@@H]2NC(=O)C(=C2[O-])C(=O)C
Specifikationer
Kemiskt namn eller material Tenuazonic acid copper(II) salt
Kvantitet 1 mg
InChI formel InChI=1S/2C10H15NO3.Cu/c2*1-4-5(2)8-9(13)7(6(3)12)10(14)11-8;/h2*5,8,13H,4H2,1-3H3,(H,11,14);/q;;+2/p-2/t2*5-,8-;/m00./s1
Formel vikt 455.1243