Läs mer
Tenuifolin, TRC
Produktkod. 17927714
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17927714 10 mg
17919834 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17927714 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT01970010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20183-47-5
Molekylformel C36 H56 O12
Molekylvikt (g/mol) 680.82
Synonym Olean-12-ene-23,28-dioic acid, 3-(β-D-glucopyranosyloxy)-2,27-dihydroxy-, (2β,3β,4α)-, Olean-12-ene-23,28-dioic acid, 3β-(β-D-glucopyranosyloxy)-2β,27-dihydroxy- (8CI), Glucopyranoside, 2β,3β,27-trihydroxyolean-12-ene-23,28-dioic acid-3, β-D- (8CI), (2β,3β,4α)-3-(β-D-Glucopyranosyloxy)-2,27-dihydroxyolean-12-ene-23,28-dioic acid, Presenegenin-3-O-glucoside, Tenuifolin, Tenuifoline, 3β-(β-D-glucopyranosyloxy)-2β,27-dihydroxy-olean-12-ene-23,28-dioic Acid, β-D-2β,3β,27-Trihydroxyolean-12-ene-23,28-dioic acid-3-glucopyranoside
IUPAC-namn (2S,3R,4S,4aR,6aR,6bR,8aS,12aS,14aR,14bR)-2-hydroxy-6b-(hydroxymethyl)-4,6a,11,11,14b-pentamethyl-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicene-4,8a-dicarboxylic acid
LEDER CC1(C)CC[C@@]2(CC[C@]3(CO)C(=CC[C@@H]4[C@@]5(C)C[C@H](O)[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@](C)([C@@H]5CC[C@@]34C)C(=O)O)[C@@H]2C1)C(=O)O
Specifikationer
Kemiskt namn eller material Tenuifolin
Kvantitet 10 mg
InChI formel InChI=1S/C36H56O12/c1-31(2)10-11-35(30(45)46)12-13-36(17-38)18(19(35)14-31)6-7-22-32(3)15-20(39)27(34(5,29(43)44)23(32)8-9-33(22,36)4)48-28-26(42)25(41)24(40)21(16-37)47-28/h6,19-28,37-42H,7-17H2,1-5H3,(H,43,44)(H,45,46)/t19-,20-,21+,22+,23+,24+,25-,26+,27-,28-,32+,33+,34-,35-,36-/m0/s1
Formel vikt 680.3772