Läs mer
Terazosin Dimer Impurity Dihydrochloride, TRC
Produktkod. 30411953
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30411953 5 mg
1 options
Produktkod. 30411953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT1050105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,2'-(Piperazin-1,4-diyl)bis-(6,7-dimethoxyquinazolin-4-amine) Dihydrochloride
CAS 104965-50-6
Molekylformel C24 H28 N8 O4 . 2 Cl H
Kvantitet 5 mg
Synonym 2,2'-(1,4-Piperazinediyl)bis[6,7-dimethoxy-4-quinazolinamine hydrochloride, 2,2'-(Piperazin-1,4-diyl)bis-(6,7-dimethoxyquinazolin-4-amine) Dihydrochloride
InChI formel InChI=1S/C24H28N8O4.2ClH/c1-33-17-9-13-15(11-19(17)35-3)27-23(29-21(13)25)31-5-7-32(8-6-31)24-28-16-12-20(36-4)18(34-2)10-14(16)22(26)30-24;;/h9-12H,5-8H2,1-4H3,(H2,25,27,29)(H2,26,28,30);2*1H
LEDER Cl.Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)c4nc(N)c5cc(OC)c(OC)cc5n4
IUPAC-namn 2-[4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine;dihydrochloride
Molekylvikt (g/mol) 565.45
Formel vikt 564.1767