Läs mer
Terazosin Hydrochloride, TRC
Produktkod. 30426197
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426197 1 g
30425471 250 mg
30416118 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426197 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT1050001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63590-64-7
Molekylformel C19 H25 N5 O4 . H Cl
Molekylvikt (g/mol) 423.89
Synonym Terazosin
IUPAC-namn [4-(4-amino-6,7-dimethoxy-quinazolin-2-yl)piperazin-1-yl]-tetrahydrofuran-2-yl-methanone;hydrochloride
LEDER Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4CCCO4
Specifikationer
Kemiskt namn eller material Terazosin, Hydrochloride
Kvantitet 1 g
InChI formel InChI=1S/C19H25N5O4.ClH/c1-26-15-10-12-13(11-16(15)27-2)21-19(22-17(12)20)24-7-5-23(6-8-24)18(25)14-4-3-9-28-14;/h10-11,14H,3-9H2,1-2H3,(H2,20,21,22);1H