missing translation for 'onlineSavingsMsg'
Läs mer

Terbinafine, TRC

Produktkod. 30135644
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135644 10 g
30133665 1 g
17923805 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30135644 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT10751310G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 91161-71-6
Molekylformel C21 H25 N
Molekylvikt (g/mol) 291.43
Synonym 1-Naphthalenemethanamine, N-[(2E)-6,6-dimethyl-2-hepten-4-yn-1-yl]-N-methyl-, 1-Naphthalenemethanamine, N-(6,6-dimethyl-2-hepten-4-ynyl)-N-methyl-, (E)-, 1-Naphthalenemethanamine, N-[(2E)-6,6-dimethyl-2-hepten-4-ynyl]-N-methyl- (9CI), N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine, Daskil, Eksifine, Fungoterbin, Lamasil, Mycoterbin, SF 86-327, TDT 067, Tebina, Terbinafine, Terbinol, Terbisil, YY 1084
IUPAC-namn (E)-N,6,6-trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine
LEDER CN(C\C=C\C#CC(C)(C)C)Cc1cccc2ccccc12

Specifications

Kemiskt namn eller material Terbinafine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C21H25N/c1-21(2,3)15-8-5-9-16-22(4)17-19-13-10-12-18-11-6-7-14-20(18)19/h5-7,9-14H,16-17H2,1-4H3/b9-5+
Formel vikt 291.1987
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.