Läs mer
Terbufoxon Sulfone, TRC
Produktkod. 30134518
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134518 25 mg
30134184 10 mg
30010203 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT10908025MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56070-15-6
Molekylformel C9 H21 O5 P S2
Molekylvikt (g/mol) 304.36
Synonym Phosphorothioic acid, S-[[(1,1-dimethylethyl)sulfonyl]methyl] O,O-diethyl ester, O,O-Diethyl S-(tert-butylsulfonyl)methyl phosphorothioate, Terbufos oxon sulfone, Terbufoxon sulfone
IUPAC-namn 2-(diethoxyphosphorylsulfanylmethylsulfonyl)-2-methylpropane
LEDER CCOP(=O)(OCC)SCS(=O)(=O)C(C)(C)C
Specifikationer
Kemiskt namn eller material Terbufos-oxon-sulfone
Kvantitet 25 mg
InChI formel InChI=1S/C9H21O5PS2/c1-6-13-15(10,14-7-2)16-8-17(11,12)9(3,4)5/h6-8H2,1-5H3
Formel vikt 304.0568