Läs mer
Terbutaline Hemisulfate Salt, TRC
Produktkod. 30005762
Change view
Kvantitet:
50 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005762 5 g
17932985 1 g
17934915 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005762 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT1097505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23031-32-5
Molekylformel 2 C12 H19 N O3 . H2 O4 S
Molekylvikt (g/mol) 548.65
Synonym Terbutaline sulfate, Bis[(1RS)-1-(3,5-dihydroxyphenyl)-2-[(1,1-dimethylethyl)amino]ethanol] sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1), 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1) (salt) (9CI), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt) (8CI), (±)-Terbutaline hemisulfate, (±)-Terbutaline sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, (±)-, sulfate (2:1) (salt), 2-(tert-Butylamino)-1-(3,5-dihydroxyphenyl)ethanol sulfate (2:1), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt), (±)-, Brethaire, Brethine, Bricanyl, Brithine, Butaliret, Monovent, Terbasmin, Terbul, Terbutalin sulfate, Terbutaline hemisulfate, Terbutaline hemisulphate, dl-Terbutaline sulfate
IUPAC-namn 5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol;sulfuric acid
LEDER CC(C)(C)NCC(O)c1cc(O)cc(O)c1.CC(C)(C)NCC(O)c2cc(O)cc(O)c2.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Terbutaline Sulfate
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/2C12H19NO3.H2O4S/c2*1-12(2,3)13-7-11(16)8-4-9(14)6-10(15)5-8;1-5(2,3)4/h2*4-6,11,13-16H,7H2,1-3H3;(H2,1,2,3,4)
Formel vikt 548.2404
Procent renhet >95%