Läs mer
Terconazole, TRC
Produktkod. 30422583
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30422583 50 mg
1 options
Produktkod. 30422583 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT11060050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Terconazole
CAS 67915-31-5
Molekylformel C26 H31 Cl2 N5 O3
Kvantitet 50 mg
Synonym Piperazine','1-[4-[[2-(2,4-dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-(1-methylethyl)-','cis-, rel-1-[4-[[(2R,4S)-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-(1-methylethyl)piperazine, (±)-Terconazole, Fungistat, Gyno-Terazol, NSC 331942, R 42470, Terazol, Terazol Cream & Suppositories, Terconazole, Tercospor, Termayazole, Triaconazole
InChI formel InChI=1S/C26H31Cl2N5O3/c1-19(2)31-9-11-32(12-10-31)21-4-6-22(7-5-21)34-14-23-15-35-26(36-23,16-33-18-29-17-30-33)24-8-3-20(27)13-25(24)28/h3-8,13,17-19,23H,9-12,14-16H2,1-2H3/t23-,26-/m0/s1
LEDER CC(C)N1CCN(CC1)c2ccc(OC[C@H]3CO[C@@](Cn4cncn4)(O3)c5ccc(Cl)cc5Cl)cc2
IUPAC-namn 1-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-propan-2-ylpiperazine
Molekylvikt (g/mol) 532.46
Formel vikt 531.1804