Läs mer
Terfuboxon, TRC
Produktkod. 30423037
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30423037 10 mg
1 options
Produktkod. 30423037 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT10907510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Terbufos-oxon
CAS 56070-14-5
Molekylformel C9 H21 O3 P S2
Kvantitet 10 mg
Synonym Phosphorothioic acid','S-[[(1,1-dimethylethyl)thio]methyl] O,O-diethyl ester, O,O-Diethyl S-(tert-butylthio)methyl phosphorothioate, Terbufos oxon, Terbufoxon
InChI formel InChI=1S/C9H21O3PS2/c1-6-11-13(10,12-7-2)15-8-14-9(3,4)5/h6-8H2,1-5H3
LEDER CCOP(=O)(OCC)SCSC(C)(C)C
IUPAC-namn 2-(diethoxyphosphorylsulfanylmethylsulfanyl)-2-methylpropane
Molekylvikt (g/mol) 272.36
Formel vikt 272.067