Läs mer
Teriflunomide, MedChemExpress
Produktkod. 16476488
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
100 mg
500 mg
Förpackningsstorlek:
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16476488 10 mg -
16462587 50 mg 50mg
16411776 500 mg 500mg
16436756 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16476488 Leverantör Medchem Express Leverantörsnummer HY15405
MedChemExpress Teriflunomide is the active metabolite of leflunomide, an approved therapy for rheumatoid arthritis. It inhibits pyrimidine synthesis and therefore potently decreases T cell and B cell proliferation.
Teriflunomide is the active metabolite of leflunomide, an approved therapy for rheumatoid arthritis. It inhibits pyrimidine synthesis and therefore potently decreases T cell and B cell proliferation.
Specifikationer
Kemiskt namn eller material Teriflunomide
Fysisk form Solid
CAS 163451-81-8
Färg Light Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C12H9F3N2O2
För användning med (applikation) COVID-19-immunoregulation
Synonym A77 1726
Löslighetsinformation DMSO : 26 mg/mL (96.22 mM; Need ultrasonic and warming)
LEDER C/C(O)=C(C#N)/C(NC1=CC=C(C(F)(F)F)C=C1)=O.[(Z)]
Molekylvikt (g/mol) 270.21
Formel vikt 270.21
Procent renhet 99.56%