Läs mer
Terlipressin Diacetate Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Terlipressin Diacetate Salt |
| CAS | 1884420-36-3 |
| Molekylformel | C56H82N16O19S2 |
| Kvantitet | 50 mg |
| Synonym | 1-Triglycyl-8-lysine Vasopressin Diacetate Salt, 130: PN: WO2010033207 SEQID: 171 claimed protein Diacetate Salt, Glycylpressin Diacetate Salt, Glypressin Diacetate Salt, Nα-Gly-Gly-Gly-8-Lys-vasopressin Diacetate Salt, Nα-Glycyl-glycyl-glycyl-[8-lysine]-vasopressin Diacetate Salt, Nα-Glycyl-glycyl-glycyl-lysine-vasopressin Diacetate Salt, Nα-Glycylglycylglycyl-vasopressin Diacetate Salt, Remestyp Diacetate Salt, Triglycyl-8-lysine-vasopressin Diacetate Salt, [N-α-Triglycyl-8-lysine]-vasopressin Diacetate Salt |
| InChI formel | InChI=1S/C52H74N16O15S2.2C2H4O2/c53-17-5-4-9-31(45(76)60-23-41(57)72)63-51(82)38-10-6-18-68(38)52(83)37-27-85-84-26-36(61-44(75)25-59-43(74)24-58-42(73)22-54)50(81)65-34(20-29-11-13-30(69)14-12-29)48(79)64-33(19-28-7-2-1-3-8-28)47(78)62-32(15-16-39(55)70)46(77)66-35(21-40(56)71)49(80)67-37;2*1-2(3)4/h1-3,7-8,11-14,31-38,69H,4-6,9-10,15-27,53-54H2,(H2,55,70)(H2,56,71)(H2,57,72)(H,58,73)(H,59,74)(H,60,76)(H,61,75)(H,62,78)(H,63,82)(H,64,79)(H,65,81)(H,66,77)(H,67,80);2*1H3,(H,3,4)/t31-,32-,33-,34-,35-,36-,37-,38-;;/m0../s1 |
| LEDER | OC(C)=O.O=C(N1CCC[C@H]1C(N[C@@H](CCCCN)C(NCC(N)=O)=O)=O)[C@@H](NC([C@H](CC(N)=O)NC([C@H](CCC(N)=O)NC([C@@H](CC2=CC=CC=C2)N3)=O)=O)=O)CSSC[C@H](NC(CNC(CNC(CN)=O)=O)=O)C(N[C@@H](CC4=CC=C(O)C=C4)C3=O)=O |
| Molekylvikt (g/mol) | 1347.48 |