Läs mer
Ternidazole Hydrochloride, TRC
Produktkod. 30009575
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30009575 25 mg
1 options
Produktkod. 30009575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT11650025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ternidazole Hydrochloride
CAS 70028-95-4
Molekylformel C7 H11 N3 O3 . Cl H
Kvantitet 25 mg
Synonym 2-methyl-5-nitro-1H-Imidazole-1-propanol hydrochloride (1:1), 1H-Imidazole-1-propanol, 2-methyl-5-nitro-, monohydrochloride (9CI), Imidazole-1-propanol, 2-methyl-5-nitro-, hydrochloride (7CI), 1-(3-Hydroxypropyl)-2-methyl-5-nitroimidazole hydrochloride
InChI formel InChI=1S/C7H11N3O3.ClH/c1-6-8-5-7(10(12)13)9(6)3-2-4-11;/h5,11H,2-4H2,1H3;1H
LEDER Cl.Cc1ncc([N+](=O)[O-])n1CCCO
IUPAC-namn 3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol;hydrochloride
Molekylvikt (g/mol) 221.64
Formel vikt 221.0567