Läs mer
tert-Butoxycarbonyl-D-proline, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | tert-Butoxycarbonyl-D-proline |
| CAS | 37784-17-1 |
| Molekylformel | C10H17NO4 |
| Kvantitet | 5 g |
| Synonym | (2R)-1,2-Pyrrolidinedicarboxylic acid 1-(1,1-dimethylethyl) ester, (R)-1,2-Pyrrolidinedicarboxylic acid, 1-(1,1-dimethylethyl) ester, (2R)-1-tert-Butoxycarbonylpyrrolidine-2-carboxylic acid, (R)-1-(tert-Butyloxycarbonyl)pyrrolidine-2-carboxylic acid, (R)-1-tert-Butoxycarbonylpyrrolidine-2-carboxylic acid, (R)-2-Carboxypyrrolidine-1-carboxylic acid tert-butyl ester, (R)-Pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 1-tert-Butoxycarbonyl-D-proline, 1-tert-Butyl (R)-pyrrolidine-1,2-dicarboxylate, N-(tert-Butoxycarbonyl)-(R)-proline, N-BOC-D-proline, N-tert-Butoxycarbonyl-D-proline, NSC 334360 |
| InChI formel | InChI=1S/C10H17NO4/c1-10(2,3)15-9(14)11-6-4-5-7(11)8(12)13/h7H,4-6H2,1-3H3,(H,12,13)/t7-/m1/s1 |
| LEDER | CC(C)(C)OC(=O)N1CCC[C@@H]1C(=O)O |
| IUPAC-namn | (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
| Molekylvikt (g/mol) | 215.25 |
| Formel vikt | 215.1158 |