Läs mer
tert-Butoxycarbonyl-D-valine, TRC
Produktkod. 30331751
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30331751 25 g
1 options
Produktkod. 30331751 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69096525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material tert-Butoxycarbonyl-D-valine
CAS 22838-58-0
Molekylformel C10H19NO4
Kvantitet 25 g
Synonym (R)-2-(tert-Butoxycarbonylamino)-3-methylbutanoic Acid, N-Boc-D-valine, N-tert-Butoxycarbonyl-D-valine, N-tert-Butyloxycarbonyl-D-valine, NSC 334316
InChI formel InChI=1S/C10H19NO4/c1-6(2)7(8(12)13)11-9(14)15-10(3,4)5/h6-7H,1-5H3,(H,11,14)(H,12,13)/t7-/m1/s1
LEDER CC(C)[C@@H](NC(=O)OC(C)(C)C)C(=O)O
IUPAC-namn (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Molekylvikt (g/mol) 217.26
Formel vikt 217.1314