Learn More
tert-Butyl 1-piperazinecarboxylate, 97%, Thermo Scientific Chemicals
Catalog Number p-3213184
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
57260-71-6
C9H19N2O2
187.26
MFCD00075265
CWXPZXBSDSIRCS-UHFFFAOYSA-O
1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
143452
tert-butyl piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CC[NH2+]CC1
57260-71-6
C9H19N2O2
187.26
MFCD00075265
CWXPZXBSDSIRCS-UHFFFAOYSA-O
1-boc-piperazine, n-boc-piperazine, 1-n-boc-piperazine, tert-butyl 1-piperazinecarboxylate, 1-tert-butoxycarbonyl piperazine, n-t-butoxycarbonylpiperazine, boc-piperazine, n-tert-butoxycarbonylpiperazine, piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 1,1-dimethylethyl ester
143452
tert-butyl piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CC[NH2+]CC1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 43°C to 47°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, V,1, 55
Formula Weight 186.25
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material tert-Butyl 1-piperazinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl 1-piperazinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .