Läs mer
tert-Butyl [(1S,2R)-1-Benzyl-2-hydroxy-3-(isobutylamino)propyl]carbamate, TRC
Produktkod. 30336956
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30336956 50 mg
1 options
Produktkod. 30336956 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69040050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material tert-Butyl [(1S,2R)-1-Benzyl-2-hydroxy-3-(isobutylamino)prop
CAS 160232-08-6
Molekylformel C19 H32 N2 O3
Kvantitet 50 mg
Synonym Carbamic acid, N-[(1S,2R)-2-hydroxy-3-[(2-methylpropyl)amino]-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, Carbamic acid, [(1S,2R)-2-hydroxy-3-[(2-methylpropyl)amino]-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester (9CI), Carbamic acid, [2-hydroxy-3-[(2-methylpropyl)amino]-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, [R-(R*,S*)]-, tert-Butyl [(1S,2R)-1-benzyl-2-hydroxy-3-(isobutylamino)propyl]carbamate
InChI formel InChI=1S/C19H32N2O3/c1-14(2)12-20-13-17(22)16(11-15-9-7-6-8-10-15)21-18(23)24-19(3,4)5/h6-10,14,16-17,20,22H,11-13H2,1-5H3,(H,21,23)/t16-,17+/m0/s1
LEDER CC(C)CNC[C@@H](O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C
IUPAC-namn tert-butyl N-[(2S,3R)-3-hydroxy-4-(2-methylpropylamino)-1-phenylbutan-2-yl]carbamate
Molekylvikt (g/mol) 336.47
Formel vikt 336.2413