Learn More
tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 291.39 |
Percent Purity | 97% |
Chemical Name or Material | tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate |