missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-(3-hydroxyphenyl)perhydro-1,4-diazepine-1-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 292.38 |
Percent Purity | 95% |
Chemical Name or Material | Tert-butyl 4-(3-hydroxyphenyl)perhydro-1, 4-diazepine-1-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'