Learn More
tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | tert-butyl 4-5-formylpyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-5-formylpyrid-2-yl piperazine-1-carboxylate, 4-5-formylpyridin-2-yl piperazine, 4-5-formylpyridin-2-yl piperazine, n1-boc protected, 1-piperazinecarboxylicacid, 4-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-formyl-2-pyridinyl-,1,1-dimethylethyl ester |
Formula Weight | 291.35 |
Percent Purity | 97% |
Chemical Name or Material | Tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate |