missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ tert-Butyl 4-aminobutyrate hydrochloride, 97%

Brand:  Alfa Aesar™ H65936.06

Additional Details : CAS Number : 58640-01-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15452669

  • 2101.85 SEK / 5g


EU Stock 1
Add to basket

Description

Description

Applications
Employed as an important intermediate for raw material for pharmaceutical.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

58640-01-0
195.687
CZVNLCOJFFYZPG-UHFFFAOYSA-N
24750478
CC(C)(C)OC(=O)CCCN.Cl
C8H18ClNO2
MFCD00270592
tert-butyl 4-aminobutanoate hydrochloride, h-gamma-abu-otbu hydrochloride, h-gamma-abu-otbu.hcl, h-gamma-abu-otbu hcl, gamma-aminobutyric acid t-butyl ester hcl, gamma-aminobutyric acid t-butyl ester hydrochloride, 4-aminobutyric acid tert-butyl ester hydrochloride, h-, a-abu-otbu.hcl, h-gaba-otbu . hcl, h-.gamma.-abu-otbu.hcl
tert-butyl 4-aminobutanoate;hydrochloride

Specifications

tert-Butyl 4-aminobutyrate hydrochloride
5g
C8H18ClNO2
MFCD00270592
CZVNLCOJFFYZPG-UHFFFAOYSA-N
tert-butyl 4-aminobutanoate;hydrochloride
24750478
Powder
White
58640-01-0
H2NCH2CH2CH2CO2C(CH3)3·HCl
tert-butyl 4-aminobutanoate hydrochloride, h-gamma-abu-otbu hydrochloride, h-gamma-abu-otbu.hcl, h-gamma-abu-otbu hcl, gamma-aminobutyric acid t-butyl ester hcl, gamma-aminobutyric acid t-butyl ester hydrochloride, 4-aminobutyric acid tert-butyl ester hydrochloride, h-, a-abu-otbu.hcl, h-gaba-otbu . hcl, h-.gamma.-abu-otbu.hcl
CC(C)(C)OC(=O)CCCN.Cl
195.687
195.69
97%
Videos
SDS