Learn More
tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.
Solubility
Soluble in chloroform, ethyl acetate, and DMSO.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.
Solubility
Soluble in chloroform, ethyl acetate, and DMSO.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Quantity | 250 mg |
Solubility Information | Soluble in chloroform,ethyl acetate,and DMSO. |
Formula Weight | 347.24 |
Percent Purity | 97% |
Chemical Name or Material | tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate |