Läs mer
tert-Butyl Fluvastatin, TRC
Produktkod. 30330548
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30330548 10 mg
1 options
Produktkod. 30330548 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69238510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,1-Dimethylethyl (3RS,5SR,6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate (Fluvastatin tert-Butyl Ester)
CAS 129332-29-2
Anteckningar om renhetsgrad HPLC
Molekylformel C28 H34 F N O4
Kvantitet 10 mg
Synonym 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, 1,1-dimethylethyl ester, (3R,5S,6E)-rel-, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, 1,1-dimethylethyl ester, [R*,S*-(E)]-(±)-, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, 1,1-dimethylethyl ester, [R*,S*-(E)]-, tert-Butyl Fluvastatin, 1,1-Dimethylethyl (3RS,5SR,6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate, Fluvastatin tert-Butyl Ester, tert-Butyl (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate
InChI formel InChI=1S/C28H34FNO4/c1-18(2)30-24-9-7-6-8-23(24)27(19-10-12-20(29)13-11-19)25(30)15-14-21(31)16-22(32)17-26(33)34-28(3,4)5/h6-15,18,21-22,31-32H,16-17H2,1-5H3/b15-14+/t21-,22-/m1/s1
LEDER CC(C)n1c(\C=C\[C@@H](O)C[C@@H](O)CC(=O)OC(C)(C)C)c(c2ccc(F)cc2)c3ccccc13
IUPAC-namn tert-butyl (E,3R,5S)-7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-3,5-dihydroxyhept-6-enoate
Molekylvikt (g/mol) 467.57
Formel vikt 467.2472