Läs mer
tert-Butyl (((Mesityl)sulfonyl)oxy)carbamate, TRC
Produktkod. 30329023
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30329023 10 g
1 options
Produktkod. 30329023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB16394010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material tert-Butyl (((Mesityl)sulfonyl)oxy)carbamate
CAS 36016-39-4
Molekylformel C14H21NO5S
Kvantitet 10 g
Synonym [(2-Methylpropan-2-yl)oxycarbonylamino] 2,4,6-Trimethylbenzenesulfonate, NSC 620029, N-tert-Butoxycarbonyl-O-(mesitylsulfonyl)hydroxylamine, 1,1-Dimethylethyl [[(2,4,6-Trimethylphenyl)sulfonyl]oxy]carbamate
InChI formel InChI=1S/C14H21NO5S/c1-9-7-10(2)12(11(3)8-9)21(17,18)20-15-13(16)19-14(4,5)6/h7-8H,1-6H3,(H,15,16)
LEDER Cc1cc(C)c(c(C)c1)S(=O)(=O)ONC(=O)OC(C)(C)C
IUPAC-namn [(2-methylpropan-2-yl)oxycarbonylamino] 2,4,6-trimethylbenzenesulfonate
Molekylvikt (g/mol) 315.39
Formel vikt 315.114