missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-7214372
Click to view available options
Quantity:
1 L
2.5 L
1634-04-4
C5H12O
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
2-methoxy-2-methylpropane
COC(C)(C)C
1634-04-4
C5H12O
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
2-methoxy-2-methylpropane
COC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -110.0°C
Density 0.7404 g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 1 L
Flash Point -28°C
Packaging Glass Bottle
Refractive Index 1.3675 to 1.3695
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Merck Index 15, 6106
Solubility Information Solubility aq. soln.: 51 g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5 g/100 g
Formula Weight 88.15
Grade ACS Reagent
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : Methyl tert-butyl ether

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'