Learn More
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate, 97%, Thermo Scientific™
Catalog Number p-2705868
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
215183-27-0
C10H14BrNO2S
292.191
MFCD03659715
CVNITIPQHIGCKI-UHFFFAOYSA-N
tert-butyl 5-bromothiophen-2-yl methyl carbamate, tert-butyl n-5-bromo-2-thienyl methyl carbamate, 5-bromo-thiophen-2-ylmethyl-carbamic acid tert-butyl ester, tert-butyl 5-bromothiophen-2-yl methylcarbamate, tert-butyl n-5-bromothiophen-2-yl methyl carbamate, n-boc-5-bromothiophen-2-methanamine, 2-boc-amino methyl-5-bromothiophene, 2-bromo-5-tert-butoxycarbonylaminomethylthiophene
2779810
tert-butyl N-[(5-bromothiophen-2-yl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC=C(S1)Br
215183-27-0
C10H14BrNO2S
292.191
MFCD03659715
CVNITIPQHIGCKI-UHFFFAOYSA-N
tert-butyl 5-bromothiophen-2-yl methyl carbamate, tert-butyl n-5-bromo-2-thienyl methyl carbamate, 5-bromo-thiophen-2-ylmethyl-carbamic acid tert-butyl ester, tert-butyl 5-bromothiophen-2-yl methylcarbamate, tert-butyl n-5-bromothiophen-2-yl methyl carbamate, n-boc-5-bromothiophen-2-methanamine, 2-boc-amino methyl-5-bromothiophene, 2-bromo-5-tert-butoxycarbonylaminomethylthiophene
2779810
tert-butyl N-[(5-bromothiophen-2-yl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC=C(S1)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 292.2
Percent Purity 97%
Chemical Name or Material Tert-butyl N-[(5-bromo-2-thienyl)methyl]carbamate