missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl phenyl carbonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.05 |
Boiling Point | 83°C |
Flash Point | 101°C (214°F) |
Linear Formula | (CH3)3COCOOC6H5 |
Quantity | 25 g |
Beilstein | 2361602 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 194.23 |
Percent Purity | 98% |
Chemical Name or Material | tert-Butyl phenyl carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'