missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
tert-Butyl Pitavastatin, TRC
Specifications
Specifications
| Kemiskt namn eller material | Pitavastatin tert-Butyl Ester |
| CAS | 586966-54-3 |
| Molekylformel | C29 H32 F N O4 |
| Kvantitet | 500 mg |
| Synonym | (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic acid 1,1-dimethylethyl ester, 1,1-Dimethylethyl (3R,5S,6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoate, Pitavastatin tert-Butyl Ester |
| InChI formel | InChI=1S/C29H32FNO4/c1-29(2,3)35-26(34)17-22(33)16-21(32)14-15-24-27(18-10-12-20(30)13-11-18)23-6-4-5-7-25(23)31-28(24)19-8-9-19/h4-7,10-15,19,21-22,32-33H,8-9,16-17H2,1-3H3/b15-14+/t21-,22-/m1/s1 |
| LEDER | CC(C)(C)OC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4 |
| IUPAC-namn | tert-butyl (E,3R,5S)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate |
| Molekylvikt (g/mol) | 477.567 |
| Formel vikt | 477.232 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?