Läs mer
tert-Butyl (R)-4,5-Diamino-5-oxopentanoate Hydrochloride, TRC
Produktkod. 30343330
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30343330 250 mg
1 options
Produktkod. 30343330 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB677320250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material tert-Butyl (R)-4,5-Diamino-5-oxopentanoate Hydrochloride
CAS 66575-26-6
Molekylformel C9H18N2O3•HCl
Kvantitet 250 mg
Synonym H-D-Glu(OtBu)-NH2 HCl, D-Glutamic Acid Gamma-tert-Butyl Ester Alpha-amide Hydrochloride
InChI formel InChI=1S/C9H18N2O3.ClH/c1-9(2,3)14-7(12)5-4-6(10)8(11)13;/h6H,4-5,10H2,1-3H3,(H2,11,13);1H/t6-;/m1./s1
LEDER O=C(OC(C)(C)C)CC[C@@H](N)C(N)=O.Cl
IUPAC-namn tert-butyl (R)-4,5-diamino-5-oxopentanoate hydrochloride hydrogen chloride
Molekylvikt (g/mol) 238.71
Formel vikt 238.12