Läs mer
tert-Butyl (Triphenylphosphoranylidene)carbamate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | tert-Butyl (Triphenylphosphoranylidene)carbamate |
| CAS | 68014-21-1 |
| Molekylformel | C23H24NO2P |
| Kvantitet | 5 g |
| Synonym | tert-Butoxycarbonylaminotriphenylphosphine, (Triphenylphosphoranylidene)carbamic Acid 1,1-Dimethylethyl Ester, N-(Triphenylphosphoranylidene)carbamic Acid 1,1-Dimethylethyl Ester |
| InChI formel | InChI=1S/C23H24NO2P/c1-23(2,3)26-22(25)24-27(19-13-7-4-8-14-19,20-15-9-5-10-16-20)21-17-11-6-12-18-21/h4-18H,1-3H3 |
| LEDER | O=C(OC(C)(C)C)N=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
| IUPAC-namn | tert-butyl (triphenyl-l5-phosphaneylidene)carbamate |
| Molekylvikt (g/mol) | 377.42 |
| Formel vikt | 377.15 |