missing translation for 'onlineSavingsMsg'
Läs mer

tert-Butyldimethylsilyl Trifluoromethanesulfonate, Sigma-Aldrich™

Product Code. 15824401
Change view
Click to view available options
Kvantitet:
25 mL
Unit Size:
25ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
15824401 25 mL 25ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15824401 Supplier Sigma Aldrich Supplier No. 8183130025
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 69739-34-0
Densitet 1.151 g/cm3 at 20°C
Kokpunkt 65°C to 67°C/13 hPa
Molekylformel CF3SO3Si(CH3)2C(CH3)3
MDL-nummer MFCD00000405
Kvantitet 25 mL
Synonym tert-Butyldimethylsilyl trifluoromethanesulfonate, Trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tert-Butyldimethylsilyl triflate, tert-Butyldimethylsilyl trifluoromethanesulfonate
Molekylvikt (g/mol) 264.34
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.