missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-80267
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7400-27-3
C4H13ClN2
124.61
MFCD00012947
DDPWVABNMBRBFI-UHFFFAOYSA-N
tert-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, monohydrochloride, tert-butylhydrazinium chloride, tert-butylhydrazine monohydrochloride, t-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, 1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, t-butylhydrazine.hcl, t-bunhnh2.hcl
81889
tert-butylhydrazine;hydrochloride
[H+].[Cl-].CC(C)(C)NN
This item is not returnable. View return policy
7400-27-3
C4H13ClN2
124.61
MFCD00012947
DDPWVABNMBRBFI-UHFFFAOYSA-N
tert-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, monohydrochloride, tert-butylhydrazinium chloride, tert-butylhydrazine monohydrochloride, t-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, 1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, t-butylhydrazine.hcl, t-bunhnh2.hcl
81889
tert-butylhydrazine;hydrochloride
[H+].[Cl-].CC(C)(C)NN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190°C to 194°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CNHNH2·HCl
Quantity 100 g
Beilstein 04, III, 1734
Fieser 12,87
Formula Weight 124.61
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material tert-Butylhydrazine hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'