missing translation for 'onlineSavingsMsg'
Learn More

TESPA1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25975-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824505

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TESPA1 is a critical component of the TCR signalosome and is essential for T cell selection and maturation through the regulation of TCR signaling during T cell development. TESPA1 encodes a protein of 458 amino acids and is highly conserved among vertebrate species. TESPA1 expression was tightly regulated during T cell development, with highest expression at the DP stage. TESPA1 deficiency resulted in failure of positive selection and substantially fewer of CD4+ and CD8+ SP cells in the thymus.
Specifications

Specifications

TESPA1
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2RU30
HSPC257, KIAA0748, TESPA1
Rabbit
IgG
20 μL
-20°C
Primary
9840
Western Blot
Unconjugated
TESPA1
Liquid
TESPA1
TESPA1 Fusion Protein Ag23218
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers