Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint Without Graduation
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
78.65 SEK - 138.86 SEK
Product Code | Brand | Height (Metric) | Diameter (Metric) | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Diameter (Metric) | Description | Price | Quantity & Availability | |||
11546944
|
Lenz Laborglasintrumente
05700034 |
150 mm | 22 mm | Without stopper |
90.68 SEK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
11556944
|
Lenz Laborglasintrumente
05700038 |
150 mm | 28 mm | Without stopper |
121.90 SEK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
11576944
|
Lenz Laborglasintrumente
05700126 |
150 mm | 18 mm | PE Stopper |
78.65 SEK
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
11596944
|
Lenz Laborglasintrumente
05700142 |
200 mm | 28 mm | PE Stopper |
138.86 SEK
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
Description
- With round bottom
- With NS socket
- Available without stopper
- With PE stopper or with hexagonal hollow glass stopper
- Made of DURAN™ tubing
Specifications
Duran Glass |