Läs mer
Testosteron (1 mg/ml i acetonitril), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Testosterone |
| CAS | 58-22-0 |
| Molekylformel | C19 H28 O2 |
| Kvantitet | 10x1 mL |
| Synonym | 17beta-Hydroxyandrost-4-en-3-one, Testosterone, Testosterone Enantate Imp. D (EP), Testosterone Propionate Imp. C (EP), Testosterone Isocaproate Imp. A (EP), Androst-4-en-3-one, 17-hydroxy-, (17β)-, Testosterone (7CI,8CI), (17β)-17-Hydroxyandrost-4-en-3-one, (+)-Testosterone, 17β-Hydroxy-Δ4-androsten-3-one, 17β-Hydroxyandrost-4-en-3-one, 17β-Hydroxyandrost-4-ene-3-one, 17β-Testosterone, 4-Androsten-3-one-17β-ol, AA 2500, Andro 100, AndroForte 5, AndroGel, Androderm, Androlin, Andronaq, Andropatch, Androst-4-en-17β-ol-3-one, Androst-4-ene-17β-ol-3-one, Andrusol, Axiron, CDB 111C, COL 1621, CP 601B, Compleo, Cristerona T, Geno-cristaux Gremy, Glowmin, Homosteron, Homosterone, Intrinsa, Mertestate, NSC 9700, Nasobol, Neotestis, Noseafix, Opterone, Oreton, Orquisteron, Perandren, Percutacrine androgenique, Primotest, Primoteston, Relibra, Striant, Synandrol F, Teslen, Testandrone, Testiculosterone, Testim, Testobase, Testoderm, Testogel, Testolent, Testolin, Testopatch, Testopel, Testopropon, Testosteroid, Testosteron, Testoviron Schering, Testoviron T, Testro AQ, Testrone, Testryl, Tostrelle, Tostrex, Viatrel, Virormone, Virosterone, Δ4-Androsten-17β-ol-3-one |
| InChI formel | InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1 |
| LEDER | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O |
| IUPAC-namn | (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimetyl-1,2,6,7,8,9,11,12,14,15,16,17-dodekahydrocyklopenta[a]fenanthren-3-one |
| Molekylvikt (g/mol) | 288.42 |
| Formel vikt | 288.2089 |
| Visa mer |