Läs mer
Testosterone 17-Isobutyrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-Oxoandrost-4-en-17beta-yl 2-Methyl-propanoate (Testosterone Isobutyrate) |
| CAS | 1169-49-9 |
| Molekylformel | C23 H34 O3 |
| Kvantitet | 100 mg |
| Synonym | 3-Oxoandrost-4-en-17beta-yl 2-Methylpropanoate, Testosterone Propionate Imp. B (EP), Testosterone Isocaproate Imp. D (EP), Testosterone isobutyrate, Androst-4-en-3-one','17-(2-methyl-1-oxopropoxy)-','(17β)-, Testosterone','isobutyrate (6CI,7CI,8CI), (17β)-17-(2-Methyl-1-oxopropoxy)androst-4-en-3-one, Isobutyric acid','ester with testosterone (8CI), 17β-(Isobutyroxy)androst-4-en-3-one, 3-Oxoandrost-4-en-17β-yl isobutyrate, Agovirin-Depot, Perandren M, Testocryst, Testosterone 17-isobutyrate, Testosterone 2-methylpropanoate, Testosterone 2-methylpropionate |
| InChI formel | InChI=1S/C23H34O3/c1-14(2)21(25)26-20-8-7-18-17-6-5-15-13-16(24)9-11-22(15,3)19(17)10-12-23(18,20)4/h13-14,17-20H,5-12H2,1-4H3/t17-,18-,19-,20-,22-,23-/m0/s1 |
| LEDER | CC(C)C(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC-namn | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 2-methylpropanoate |
| Molekylvikt (g/mol) | 358.51 |
| Formel vikt | 358.2508 |