Läs mer
Testosterone Isocaproate 1.0 mg/ml in Acetonitrile, LoGiCal
Produktkod. 30462134
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30462134 1 ml
1 options
Produktkod. 30462134 Leverantör LoGiCal Leverantörsnummer LGCAMP0863.0011
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Testosterone isocaproate
CAS 15262-86-9
Analyt- eller komponentnamn Testosterone isocaproate
Frakt skick Ice pack (-20°C)
Molekylformel C25 H38 O3
Certifikat och efterlevnad ISO 17034
Synonym Testosterone, 4-methylvalerate (6CI,8CI), Valeric acid, 4-methyl-, ester with testosterone (8CI), NSC 26641, Testosterone 17-isocaproate, Testosterone 4-methylpentanoate, Testosterone isocaproate, Androst-4-en-3-one, 17-[(4-methyl-1-oxopentyl)oxy]-, (17beta)-, (17beta)-17-[(4-Methyl-1-oxopentyl)oxy]androst-4-en-3-one, 3-Oxoandrost-4-en-17beta-yl isocaproate
InChI formel InChI=1S/C25H38O3/c1-16(2)5-10-23(27)28-22-9-8-20-19-7-6-17-15-18(26)11-13-24(17,3)21(19)12-14-25(20,22)4/h15-16,19-22H,5-14H2,1-4H3/t19-,20-,21-,22-,24-,25-/m0/s1
Lösningstyp Single Solution
LEDER CC(C)CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 4-methylpentanoate
Lösningsmedel eller matris Acetonitrile
Industry Type Clinical Testing
Molekylvikt (g/mol) 386.57
Kvantitet 1 ml
Formel vikt 386.2821 g/mol