Läs mer
Tetra(hydroxyethyl)rutoside (>85%), TRC
Produktkod. 30415078
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30415078 100 mg
1 options
Produktkod. 30415078 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT294310100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3',4',5,7-Tetra(hydroxyethyl)rutoside
CAS 6980-20-7
Molekylformel C35H46O20
Kvantitet 100 mg
Synonym 3',4',5,7-Tetrakis-O-(2-hydroxyethyl)-rutin, beta-D-2-[3,4-Bis(2-hydroxyethoxy)phenyl]-5,7-bis(2-hydroxyethoxy)-4-oxo-4H-1-benzopyran-3-yl O-(6-Deoxy-alpha-L-mannopyranosyl)-glucopyranoside, 3',4',5,7-Tetra-O-(beta-hydroxyethyl)rutoside, 5,7,3',4'-Tetra-O-hydroxyethylrutoside, 5,7,3',4'-Tetra-O-beta-hydroxyethylrutoside, 5,7,3',4'-Tetrakis-O-(hydroxyethyl)rutin, O-3',4',5,7-Tetrakis(beta-hydroxyethyl)rutoside, Tetra-O-(beta-hydroxyethyl)rutoside, TetraHER, Tetrakis-O-(2-hydroxyethyl)rutin, Z 1200
InChI formel InChI=1S/C35H46O20/c1-16-25(40)28(43)30(45)34(52-16)51-15-23-26(41)29(44)31(46)35(54-23)55-33-27(42)24-21(50-11-7-39)13-18(47-8-4-36)14-22(24)53-32(33)17-2-3-19(48-9-5-37)20(12-17)49-10-6-38/h2-3,12-14,16,23,25-26,28-31,34-41,43-46H,4-11,15H2,1H3/t16-,23+,25-,26+,28+,29-,30+,31+,34+,35-/m0/s1
LEDER C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(OCCO)cc(OCCO)c4C3=O)c5ccc(OCCO)c(OCCO)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O
IUPAC-namn 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5,7-bis(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
Molekylvikt (g/mol) 786.7
Formel vikt 786.2582