missing translation for 'onlineSavingsMsg'
Läs mer

Tetra-n-butylammonium Iodide, Sigma-Aldrich™

Product Code. 15891691
Change view
Click to view available options
Kvantitet:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
15891691 250 g 250g
15881691 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15891691 Supplier Sigma Aldrich Supplier No. 8221520250
 Begränsad artikel
Beräknad leverans: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311-28-4

Specifications

Smältpunkt 143°C to 146°C
MDL-nummer MFCD00011636
Kvantitet 250 g
Synonym Tetra-n-butylammonium iodide, Tetra-n-butylammonium iodide
Molekylvikt (g/mol) 369.37
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.