Läs mer
Tetra-O-pivaloyl-Alpha-D-glucopyranosyl Bromide, TRC
Produktkod. 30452548
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30452548 1 g
1 options
Produktkod. 30452548 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3063601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetra-O-pivaloyl-alpha-D-glucopyranosyl Bromide
CAS 81058-27-7
Molekylformel C26H43BrO9
Kvantitet 1 g
Synonym 2,3,4,6-Tetrakis(2,2-dimethylpropanoate) alpha-D-Glucopyranosyl Bromide, 2,3,4,6-Tetra-O-pivaloyl-alpha-D-glucopyranosyl bromide
InChI formel InChI=1S/C26H43BrO9/c1-23(2,3)19(28)32-13-14-15(34-20(29)24(4,5)6)16(35-21(30)25(7,8)9)17(18(27)33-14)36-22(31)26(10,11)12/h14-18H,13H2,1-12H3/t14-,15-,16+,17-,18+/m1/s1
LEDER CC(C)(C)C(=O)OC[C@H]1O[C@H](Br)[C@H](OC(=O)C(C)(C)C)[C@@H](OC(=O)C(C)(C)C)[C@@H]1OC(=O)C(C)(C)C
IUPAC-namn [(2R,3R,4S,5R,6R)-6-bromo-3,4,5-tris(2,2-dimethylpropanoyloxy)oxan-2-yl]methyl 2,2-dimethylpropanoate
Molekylvikt (g/mol) 579.52
Formel vikt 578.209