Läs mer
Tetra-salicyclic Acid, TRC
Produktkod. 30456866
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30456866 50 mg
1 options
Produktkod. 30456866 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30644550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Salicylsalicylsalicylsalicylic Acid
CAS 85531-18-6
Molekylformel C28 H18 O9
Kvantitet 50 mg
Synonym Tetra-salicyclic Acid, Benzoic acid','2-[(2-hydroxybenzoyl)oxy]-','2-[(2-carboxyphenoxy)carbonyl]phenyl ester, Tetrasalicylic Acid
InChI formel InChI=1S/C28H18O9/c29-21-13-5-1-9-17(21)26(32)36-23-15-7-3-11-19(23)28(34)37-24-16-8-4-12-20(24)27(33)35-22-14-6-2-10-18(22)25(30)31/h1-16,29H,(H,30,31)
LEDER O=C(O)C=1C=CC=CC1OC(=O)C=2C=CC=CC2OC(=O)C=3C=CC=CC3OC(=O)C=4C=CC=CC4O
IUPAC-namn 2-[2-[2-(2-hydroxybenzoyl)oxybenzoyl]oxybenzoyl]oxybenzoic acid
Molekylvikt (g/mol) 498.437
Formel vikt 498.095