missing translation for 'onlineSavingsMsg'
Learn More

Tetraamminepalladium(II) hydrogen carbonate, Thermo Scientific Chemicals

Catalog Number p-7059652
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
134620-00-1
C2H14N4O6Pd
296.576
MFCD09953449
QYYPTZGPXKUKRM-UHFFFAOYSA-L
palladium 2+ , tetraammine-, sp-4-1-, carbonate 1:2, azane; hydrogen carbonate; palladium 2+, tetraamminepalladium ii hydrogencarbonate, ammonia; hydrogen carbonate; palladium 2+, palladium 2+ tetraamine dibicarbonate, tetraammine palladium ii hydrogen carbonate, palladium 2+ ion tetraamine dibicarbonate
153931
azane;hydrogen carbonate;palladium(2+)
C(=O)(O)[O-].C(=O)(O)[O-].N.N.N.N.[Pd+2]
This item is not returnable. View return policy
134620-00-1
C2H14N4O6Pd
296.576
MFCD09953449
QYYPTZGPXKUKRM-UHFFFAOYSA-L
palladium 2+ , tetraammine-, sp-4-1-, carbonate 1:2, azane; hydrogen carbonate; palladium 2+, tetraamminepalladium ii hydrogencarbonate, ammonia; hydrogen carbonate; palladium 2+, palladium 2+ tetraamine dibicarbonate, tetraammine palladium ii hydrogen carbonate, palladium 2+ ion tetraamine dibicarbonate
153931
azane;hydrogen carbonate;palladium(2+)
C(=O)(O)[O-].C(=O)(O)[O-].N.N.N.N.[Pd+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 2.04 g/mL
Linear Formula [Pd(NH3)4](HCO3)2
Quantity 1 g
UN Number UN3077
Formula Weight 296.58
Physical Form Crystalline
Chemical Name or Material Tetraamminepalladium (II) hydrogen carbonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'