missing translation for 'onlineSavingsMsg'
Läs mer

Tetrabenazine, Mikromol™

Produktkod. 17971076
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17971076 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17971076 Leverantör Mikromol Leverantörsnummer MM1633.01

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tetrabenazine
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Tetrabenazine
CAS 58-46-8
Analyt- eller komponentnamn Tetrabenazine
Frakt skick Room Temperature
Molekylformel C19 H27 N O3
Certifikat och efterlevnad ISO 17034
Synonym 2H-Benzo[a]quinolizin-2-one, 1,3,4,6,7,11b-hexahydro-3-isobutyl-9,10-dimethoxy- (6CI,8CI), 2H-Benzo[a]quinolizin-2-one, 1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-, cis-, NSC 169886, NSC 172187, Ro 1-9569, Rubigen, Tetrabenazine, Xenazine, cis-2-Oxo-3-isobutyl-9,10-dimethoxy-1,2,3,4,6,7-hexahydro-11bH-benzo[a]quinolizine, 2H-Benzo[a]quinolizin-2-one, 1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-, (3R,11bR)-rel-, (+/-)-Tetrabenazine
InChI formel InChI=1S/C19H27NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-9,12,14,16H,5-7,10-11H2,1-4H3/t14-,16-/m0/s1
LEDER COc1cc2CCN3C[C@H](CC(C)C)C(=O)C[C@H]3c2cc1OC
IUPAC-namn (3S,11bS)-9,10-dimethoxy-3-(2-methylpropyl)-1,3,4,6,7,11b-hexahydrobenzo[a]quinolizin-2-one
Molekylvikt (g/mol) 317.42
Kvantitet 100 mg
Formel vikt 317.1991 g/mol
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.