missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromo-p-benzoquinone, 95%, Thermo Scientific™

Catalog Number p-3213216
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
488-48-2
C6Br4O2
423.68
MFCD00013785
LWHDQPLUIFIFFT-UHFFFAOYSA-N
bromanil, tetrabromo-1,4-benzoquinone, bromanyl, p-bromanil, tetrabromoquinone, tetrabromo-p-benzoquinone, p-bromoanil, tetrabromo-p-quinone, tetrabromobenzoquinone, p-quinone, tetrabromo
68101
BrC1=C(Br)C(=O)C(Br)=C(Br)C1=O
This item is not returnable. View return policy
488-48-2
C6Br4O2
423.68
MFCD00013785
LWHDQPLUIFIFFT-UHFFFAOYSA-N
bromanil, tetrabromo-1,4-benzoquinone, bromanyl, p-bromanil, tetrabromoquinone, tetrabromo-p-benzoquinone, p-bromoanil, tetrabromo-p-quinone, tetrabromobenzoquinone, p-quinone, tetrabromo
68101
BrC1=C(Br)C(=O)C(Br)=C(Br)C1=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 290°C to 297°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula C6Br4(=O)2
Quantity 1 g
IUPAC Name 2,3,5,6-tetrabromocyclohexa-2,5-diene-1,4-dione
Formula Weight 423.68
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromo-p-benzoquinone
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabromo-p-benzoquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'